Order Entry
Belgium
ContactUsLinkComponent
Iodine 0.02365 mol/l (0.0473N) ASTM D 1510 volumetric solution
Iodine 0.02365 mol/l (0.0473N) ASTM D 1510 volumetric solution
Catalog # APPC182915.1612
Supplier:  PanReac AppliChem
CAS Number:  
Iodine 0.02365 mol/l (0.0473N) ASTM D 1510 volumetric solution
Catalog # APPC182915.1612
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Titer at 20°C:
    0.999 - 1.001

Specification Test Results

About this item

 :
 :253.81 g/mol
Storage Temperature:Ambient
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355