Order Entry
Belgium
Orders LinkContactUsLinkComponent
Triethylammonium bicarbonate buffer, 1 M, volatile buffer for HPLC, Fluka™

Triethylammonium bicarbonate buffer, 1 M, volatile buffer for HPLC, Fluka™

Supplier:   Honeywell Chemicals
Triethylammonium bicarbonate buffer, 1 M, volatile buffer for HPLC, Fluka™
Certificates
Triethylammonium bicarbonate buffer, 1 M, volatile buffer for HPLC, Fluka™

2 Options Available Below

Product Details & Documents

Triethylammonium bicarbonate buffers are volatile buffer for HPLC.

  • Non-volatile matter: max. 0,05 %
  • pH (20 °C): 8,4 to 8,6
  • Absorbance at 260 nm: Max. 0,04
  • Absorbance at 280 nm: Max. 0,04
Product Family Options

Product Information

  • pH value
    Pack type
    Pk
    Availability
    Price
  • 17902-100ML
    Triethylammonium bicarbonate buffer, 1 M
    Triethylammonium bicarbonate buffer, 1 M
    8,4 - 8,6
    Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 126,00 €
    126,00 €
    1 * 100 mL
       126,00 €

  • 17902-500ML
    Triethylammonium bicarbonate buffer, 1 M
    Triethylammonium bicarbonate buffer, 1 M
    8,4 - 8,6
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 464,00 €
    464,00 €
    1 * 500 mL
       464,00 €

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.