Phosphate buffer solution pH 7,00 (20 °C), Fluka™
Phosphate buffer solution, 7,00, ±0,02, Plastic bottle, 1 L
Catalog # 73173-1L
Supplier: Honeywell Chemicals
Phosphate buffer solution pH 7,00 (20 °C), Fluka™
Catalog # 73173-1L
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Potassium dihydrogen phosphate / di-sodium hydrogen phosphate
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Description:Phosphate buffer solution
- pH accuracy:±0,02
- pH value:7,00



