
Magnesium ≥99% (by complexometric analysis), filings, pure
APPC141945.0616
:
- Pk:25 kg
- Seq:4283
- Assay (Complexometric):99%
- Insoluble Matter in HCl:Passes Test
- Al:≤0.05%
- Cu:≤0.005%
- Fe:≤0.05%
- Mn:≤0.1%
- Ni:≤0.005%
- PB:≤0.005%
- Si:≤0.1%
- Zn:≤0.005%
:
:24.3 g/mol
:1107 °C
:651 °C
:Ambient
:231-104-6
:1869
:4.1,III
:GHS02
:7439-95-4
:Mg
:MFCD00085308