Order Entry
Belgium
ContactUsLinkComponent
Silica gel, Pore size: 60 Å ≥99%, powder 200 - 425 mesh, high purity (Davisil Grade 633), Supelco®
Silica gel, Pore size: 60 Å ≥99%, powder 200 - 425 mesh, high purity (Davisil Grade 633), Supelco®
Supplier:  Merck
Certificates
undefined
Silica gel, Pore size: 60 Å ≥99%, powder 200 - 425 mesh, high purity (Davisil Grade 633), Supelco®
Supplier:  Merck

Specification Test Results

  • Appearance (Colour):
    White
  • Appearance (Form):
    Powder
  • Particle Size: 200 - 425 Mesh:
    Conforms
  • Assay:
    ≥99%

Specification Test Results

Frequently Bought Together