Orders LinkContactUsLinkComponent
Adapters, PTFE, GL thread, BOLA
Adapters, PTFE, GL thread, BOLA
Supplier:   Bohlender
Adapters, PTFE, GL thread, BOLA
Supplier:  Bohlender
Certificates
Adapters, PTFE, GL thread, BOLA
Adapters, PTFE, GL thread, BOLA
2 Options Available Below
Product Details & Documents
Adaptor made of glass-fibre reinforced PTFE.
  • Transition from pump thread M20x1,5 to GL thread
  • Pressure resistant connection of maximum 10 bar of hardwalled tubing
  • Universal chemical resistance
  • Temperature resistant from –200 to +250 °C
Product Family Options
Product Information
  • Thread
    Hole Ø
    Availability
    Price
  • GL 14
    3,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,80 €
    82,80 €
    1 * 1 ST
       82,80 €
  • GL 18
    10,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,50 €
    86,50 €
    1 * 1 ST
       86,50 €