0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Standards
Environmental Analysis Standards
Base/Neutrals 1 - WP, (CRM), Supelco®
Base/Neutrals 1 - WP, (CRM), Supelco®
Catalog # QC1173-2ML
Supplier:
Merck
CAS Number:
Expand image
Base/Neutrals 1 - WP, (CRM), Supelco®
Catalog # QC1173-2ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
2 mL
Description:
Base/Neutrals 1 - WP
Type:
Certified reference material
Specifications
About this item
Product Details
Product Details