Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Trace metals - sewage sludges, (CRM), Supelco®
Trace metals - sewage sludges, (CRM), Supelco®
Trace metals - sewage sludges, (CRM), Supelco®
Catalog # CRM031-40G
Supplier:
Merck
CAS Number:
Certificates
Expand image
Trace metals - sewage sludges, (CRM), Supelco®
Catalog # CRM031-40G
Supplier:
Merck
CAS Number:
Certificates
Specifications
Pk:
40 g
Description:
Trace Metals - Sewage Sludge 3
Pk Info:
Pkg of 40 g (dry)
Specifications
About this item
Product Details
Documentation
Product Details
Documentation