Supelco® Organic Reference Standard, HDI-DBA Oligomers Standard
CRM40589
: Merck
:
- Concentration:In methanol (varied conc.)
- Pk:1 mL
- Standards:Certified reference material
- Description:HDI-DBA oligomers standard
- Storage conditions:2…8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Synonym(s):HDI adducts; Hexamethylene diisocyanate dibutylamine oligomers standard
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1×1 ml
- GHS signal: GHS02, GHS06, GHS08
: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311