Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, 48 Component Indoor Air Standard
Supelco® Organic Reference Standard, 48 Component Indoor Air Standard
Catalog # 40353-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, 48 Component Indoor Air Standard
Catalog # 40353-U
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    Certified reference material, 1000 mug/ml each component in methanol: water (19:1), ampule of 1 ml
  • Description:
    48 Component Indoor Air Standard
  • Storage conditions:
    –10...–25 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H340 - H350 - H370 - H412
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Pk Info:
    Ampoule of 1 mL

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08, GHS09
Caution: P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311