ContactUsLinkComponent
Chemistry vacuum pumps and systems with valve control, dual application
Chemistry vacuum pumps and systems with valve control, dual application
Supplier:  VACUUBRAND
Certificates
undefined
Chemistry vacuum pumps and systems with valve control, dual application
Supplier:  VACUUBRAND
Specifications
  • Catalog No:
  • 131-0040
  • 131-0026
  • 131-0037
  • 131-0043
  • 131-0020
  • 131-0023
  • Model:
  • PC 611 select (CEE-plug)
  • PC 520 select (CEE-plug)
  • PC 610 select (CEE-plug)
  • PC 620 select (CEE-plug)
  • PC 510 select (CEE-plug)
  • PC 511 select (CEE-plug)
  • No. of stages:
  • 3
  • 2
  • 3
  • 3
  • 2
  • 2
  • Flow rate:
  • 57 l/min
  • 34 l/min
  • 57 l/min
  • 57 l/min
  • 34 l/min
  • 34 l/min
  • Vacuum:
  • 3,0 mbar (2,2 torr), with gas ballast
  • 12,0 mbar (9,0 torr), with gas ballast
  • 3,0 mbar (2,2 torr), with gas ballast
  • 3,0 mbar (2,2 torr), with gas ballast
  • 12,0 mbar (9,0 torr), with gas ballast
  • 12,0 mbar (9,0 torr), with gas ballast
  • Ultimate vacuum:
  • 1,5 mbar (1,1 torr)
  • 7,0 mbar (5,0 torr)
  • 1,5 mbar (1,1 torr)
  • 1,5 mbar (1,1 torr)
  • 7,0 mbar (5,0 torr)
  • 7,0 mbar (5,0 torr)