5-Carboxy-2',7'-dichlorofluorescein ≥98% (by TLC) fluorescent pH indicator
Catalog # 51017.
Supplier: Biotium
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:445 g/mol
Storage Temperature:Refrigerator
:111843-78-8
Formula:C₂₁H₁₀Cl₂O₇