5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA) ≥95% (by TLC) fluorescent pH indicator
Catalog # 90040.
Supplier: Biotium
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:626.35 g/mol
Storage Temperature:Freezer
CAS Number:147265-60-9
Formula:C₂₉H₁₇Cl₂NO₁₁