Supelco® Cerilliant® Organic Reference Standard, 4-Hydroxy-3-methoxyphenyl-D₃-acetic-D₂ Acid solution
N° de catalogue H-092-1ML
Fournisseur: Merck
Numéro CAS:
Spécifications
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:53587-32-9
- Description:4-Hydroxy-3-methoxyphenyl-D₃-acetic-D₂ Acid solution
- Storage conditions:–70 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:DICE
- M-Clarity quality level:MQ300
- Synonym(s):4-Hydroxy-3-methoxyphenyl-d₃-acetic-d₂ Acid (HVA-d₅); HVA-D₅ solution
- Molecular weight:187,20
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
Spécifications
À propos de cet article
- GHS signal: GHS02, GHS06, GHS08
Attention: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311