Sulphorhodamine 101-PEO₈-propionic acid succinimydyl ester amine-reactive fluorescent dye
Catalog # 90099.
Supplier: Biotium
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:1127.28 g/mol
Storage Temperature:Freezer
:60311-02-6
Formula:C₅₄H₇₀N₄O₁₈S₂