Order Entry
Belgium
ContactUsLinkComponent
6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE) ≥95% (by TLC) amine-reactive fluorescent dye
6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE) ≥95% (by TLC) amine-reactive fluorescent dye
Catalog # 90033.
Supplier:  Biotium
CAS Number:  
undefined
6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE) ≥95% (by TLC) amine-reactive fluorescent dye
Catalog # 90033.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Carboxyrhodamine 6G succinimidyl ester, 6-isomer, 6-CR6G, NHS ester, 6-Carboxyrhodamine 6G, succinimidyl ester
 :
 :555.59 g/mol
Storage Temperature:Freezer
CAS Number:216699-37-5
Formula:C₃₁H₂₉N₃O₇