Order Entry
Belgium
ContactUsLinkComponent
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥99% (by TLC) amine-reactive fluorescent dye
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥99% (by TLC) amine-reactive fluorescent dye
Catalog # 90029.
Supplier:  Biotium
CAS Number:  
undefined
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥99% (by TLC) amine-reactive fluorescent dye
Catalog # 90029.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :473.39 g/mol
Storage Temperature:Freezer
CAS Number:92557-80-7
Formula:C₂₅H₁₅NO₉
MDL Number:MFCD00077322