Order Entry
Belgium
ContactUsLinkComponent
Tetramethylrhodamine 5-isothiocyanate chloride ≥90% (by TLC) amine-reactive fluorescent dye
Tetramethylrhodamine 5-isothiocyanate chloride ≥90% (by TLC) amine-reactive fluorescent dye
Catalog # 90018.
Supplier:  Biotium
CAS Number:  
Tetramethylrhodamine 5-isothiocyanate chloride ≥90% (by TLC) amine-reactive fluorescent dye
Catalog # 90018.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :479.98 g/mol
Storage Temperature:Freezer
Formula:C₂₅H₂₂ClN₃O₃S