Order Entry
Belgium
ContactUsLinkComponent
 
Transition metal mix 2 for ICP, 8 elements, TraceCERT®, Supelco®
  96442-100ML
 :  Merck
 :  
undefined
Transition metal mix 2 for ICP, 8 elements, TraceCERT®, Supelco®
  96442-100ML
 :  Merck
 :  

 

  • Concentration:
    100 mg/L
  • Matrix:
    2% HNO₃/1% HF
  • Pk:
    100 mL
  • Description:
    Transition metal mix 2 for ICP
  • Environmentally Preferable:

 

 

100 mg/l each: Ti, Zr, Hf, Nb, Ta, Mo, W and Re in 2% nitric acid and 1% hydrofluoric acid

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034