Order Entry
Belgium
ContactUsLinkComponent
Transition metal mix 3 for ICP, 7 elements, TraceCERT®, Supelco®
Transition metal mix 3 for ICP, 7 elements, TraceCERT®, Supelco®
Catalog # 93468-100ML
Supplier:  Merck
CAS Number:  
undefined
Transition metal mix 3 for ICP, 7 elements, TraceCERT®, Supelco®
Catalog # 93468-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    10% HCl
  • Pk:
    100 mL
  • Description:
    Transition metal mix 3 for ICP

Specifications

About this item

100 mg/l each: Ru, Rh, Pd, Os, Ir, Pt and Au in 10% hydrochloric acid

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034