Order Entry
Belgium
ContactUsLinkComponent
Rare earth element mix for ICP, 16 elements, TraceCERT®, Supelco®
Rare earth element mix for ICP, 16 elements, TraceCERT®, Supelco®
Catalog # 67349-100ML
Supplier:  Merck
CAS Number:  
undefined
Rare earth element mix for ICP, 16 elements, TraceCERT®, Supelco®
Catalog # 67349-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    50 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Description:
    Rare earth element mix for ICP

Specifications

About this item

50 mg/l each: Sc, Y, La, Ce, Pr, Nd, Sm, Eu, Gd, Tb, Dy, Ho, Er, Tm, Yb and Lu in 2% nitric acid

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034