Order Entry
Belgium
ContactUsLinkComponent
 
Metalloid and non-metal mix for ICP, 10 elements, TraceCERT®, Supelco®
Catalog # 55263-100ML
Supplier:  Merck
CAS Number:  
undefined
Metalloid and non-metal mix for ICP, 10 elements, TraceCERT®, Supelco®
Catalog # 55263-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    10% HNO₃/tr HF
  • Pk:
    100 mL
  • Description:
    Metalloid and non-metal mix for ICP

Specifications

About this item

100 mg/l: B, Si, P, S, Ge, As, Se, Sn, Sb and Te in 10% nitric acid and ≤0.5% hydrofluoric acid

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034