Periodic table mix 2 for ICP, 17 elements, TraceCERT®, Supelco®
Catalog # 41135-100ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:5% HCl/tr HF
- Pk:100 mL
- Description:Periodic table mix 2 for ICP
- Environmentally Preferable:
Specifications
About this item
10 mg/L: Au, Ge, Hf, Ir, Mo, Nb, Pd, Pt, Re, Rh, Ru, Sb, Sn, Ta, Ti, W, Zr in 5%hydrochloric acid and 1% hydrofluoric acid (contains HNO₃ traces)
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034