Order Entry
Belgium
ContactUsLinkComponent
Transition metal mix 1 for ICP, 10 elements, TraceCERT®, Supelco®
Transition metal mix 1 for ICP, 10 elements, TraceCERT®, Supelco®
Catalog # 04330-100ML
Supplier:  Merck
CAS Number:  
Transition metal mix 1 for ICP, 10 elements, TraceCERT®, Supelco®
Catalog # 04330-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Description:
    Transition metal mix 1 for ICP

Specifications

About this item

100 mg/l each: V, Cr, Mn, Fe, Co, Ni, Cu, Zn, Ag and Cd in 2% nitric acid

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034