Order Entry
Belgium
ContactUsLinkComponent
5(6)-Carboxyrhodamine 110 hydrochloride ≥90% (by HPLC) amine-reactive fluorescent dye
5(6)-Carboxyrhodamine 110 hydrochloride ≥90% (by HPLC) amine-reactive fluorescent dye
Catalog # 90000.
Supplier:  Biotium
CAS Number:  
undefined
5(6)-Carboxyrhodamine 110 hydrochloride ≥90% (by HPLC) amine-reactive fluorescent dye
Catalog # 90000.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :410.81 g/mol
Storage Temperature:Refrigerator
Formula:C₂₂H₁₉ClN₂O₅