0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, nimesulide, (CRM), Supelco®
Pharmaceutical secondary standard, nimesulide, (CRM), Supelco®
PHR1336-1G
:
Merck
:
Pharmaceutical secondary standard, nimesulide, (CRM), Supelco®
PHR1336-1G
:
Merck
:
Pk:
1 g
CAS No.:
51803-78-2
Description:
Nimesulide
Description:
Nimesulide