Order Entry
Belgium
ContactUsLinkComponent
 
Pharmaceutical secondary standard, ibuprofen, (CRM), Supelco®
Catalog # PHR1004-1G
Supplier:  Merck
CAS Number:  
undefined
Pharmaceutical secondary standard, ibuprofen, (CRM), Supelco®
Catalog # PHR1004-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • CAS No.:
    15687-27-1
  • Description:
    Ibuprofen
  • Formula:
    C₁₃H₁₈O₂
  • Description:
    Ibuprofen
  • Form:
    Neat

Specifications

About this item

  • Traceable to BP 539
  • Traceable to PhEur I0020000
  • Traceable to USP 1335508
Certifications: This product was designed, produced and verified for accuracy and stability in accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO GUIDES 34:2009 (AClass Cert AR-1470).