0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, Eugenol, (CRM), Supelco®
Pharmaceutical secondary standard, Eugenol, (CRM), Supelco®
PHR1269-1G
:
Merck
:
Pharmaceutical secondary standard, Eugenol, (CRM), Supelco®
PHR1269-1G
:
Merck
:
Pk:
1 g
CAS No.:
97-53-0
Description:
Eugenol
Description:
Eugenol