0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, Acyclovir, (CRM), Supelco®
Pharmaceutical secondary standard, Acyclovir, (CRM), Supelco®
Catalog # PHR1254-1G
Supplier:
Merck
CAS Number:
Certificates
Expand image
Pharmaceutical secondary standard, Acyclovir, (CRM), Supelco®
Catalog # PHR1254-1G
Supplier:
Merck
CAS Number:
Certificates
Specifications
Pk:
1 g
CAS No.:
59277-89-3
Description:
Acyclovir
Description:
Acyclovir
Specifications
About this item
Product Details
Documentation
Product Details
Documentation