Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Acetaminophen Impurity G
Supelco® Organic Reference Standard, Acetaminophen Impurity G
  PHR1972-100MG
 :  Merck
 :  
Supelco® Organic Reference Standard, Acetaminophen Impurity G
  PHR1972-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Pharmaceutical Secondary Standard; Certified Reference Material
  • Description:
    Acetaminophen impurity G
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    1-(4-hydroxyphenyl)ethanone oxime; 1-(4-hydroxyphenyl)ethanone oxime
  • Formula:
    C8H9NO2
  • Molecular weight:
    151,16