Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, 2-Phenoxyethanol
Supelco® Organic Reference Standard, 2-Phenoxyethanol
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, 2-Phenoxyethanol
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    122-99-6
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H318 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002857
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    Ethylene glycol monophenyl ether; PHE; PHE-G; PHE-S; PhG; Phenylglycol
  • Melting point:
    11 - 13 °C
  • Formula:
    C₆H₅OCH₂CH₂OH
  • Molecular weight:
    138,16
  • Density:
    1,107 g/ml at 20 °C (lit.)
  • Flash point:
    126 °C - Closed cup
  • Boiling point:
    244 - 246 °C

Specifications

Related Information

Frequently Bought Together