Supelco® TraceCERT® Organic Reference Standard, Pyrocatechol
Pyrocatechol, Certified reference material, TraceCERT®, 100 mg
Catalog # 50550-100MG
Supplier: Merck
Supelco® TraceCERT® Organic Reference Standard, Pyrocatechol
Catalog # 50550-100MG
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
- GHS signal: GHS06, GHS08, GHS05
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P202 - P261 - P280 - P301 + P310 - P302 + P352 + P312 - P305 + P351 + P338
Specifications
Specifications
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- Description:Pyrocatechol
- Boiling point:245 °C (lit.)
- Hazard class description:H301 + H311 - H315 - H317 - H318 - H341 - H350
- MDL No.:MFCD00002188
- Synonym(s):1,2-Benzenediol; 1,2-Dihydroxybenzene; 2-Hydroxyphenol; Catechol
- Molecular weight:110,11
- CAS No.:120-80-9
- Cat. No.:50550-100MG
- M-Clarity quality level:MQ300
- Formula:C₆H₄-1,2-(OH)₂
- Storage conditions:15...25 °C
- Flash point:127 °C - Closed cup
- Melting point:100 - 103 °C (lit.)
- Shipping conditions:STD



