Organic reference standard, neat, Bisphenol A, (CRM), TraceCERT®, Supelco®
Catalog # 42088-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- CAS No.:80-05-7
- Description:Bisphenol A
- MDL No.:MFCD00002366
- M-Clarity quality level:MQ300
- Description:Bisphenol A
Specifications
About this item
Certified Reference material suitable for HPLC and GC analysis. Produced and certified in accordance with ISO/IEC 17025 and ISO 17034.