Order Entry
Belgium
ContactUsLinkComponent
Food standards, Deoxynivalenol solution, (CRM), Supelco®
Food standards, Deoxynivalenol solution, (CRM), Supelco®
Catalog # CRM46911
Supplier:  Merck
CAS Number:  
undefined
Food standards, Deoxynivalenol solution, (CRM), Supelco®
Catalog # CRM46911
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    200 µg/ml in ethyl acetate: methanol (95:5)
  • Pk:
    1
  • CAS No.:
    51481-10-8
  • Description:
    Deoxynivalenol solution
  • Description:
    Deoxynivalenol solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item