Order Entry
Belgium
ContactUsLinkComponent
Food standards, T-2 Toxin, neat, Supelco®
Food standards, T-2 Toxin, neat, Supelco®
Catalog # 33947-5MG
Supplier:  Merck
CAS Number:  
undefined
Food standards, T-2 Toxin, neat, Supelco®
Catalog # 33947-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pk:
    5 mg
  • CAS No.:
    21259-20-1
  • Description:
    T-2 Toxin
  • M-Clarity quality level:
    MQ200
  • Description:
    T-2 Toxin

Specifications

About this item

T-2 Toxin is found to be a potent trichothecene mycotoxin produced by the genus fusarium fungi in feedstuffs and cereal grains.

Caution: Storage temperature: Freezer −20 °C