Order Entry
Belgium
Orders LinkContactUsLinkComponent
Papain

Papain

—, 5 kg
Catalog # 1.07144.5000
Supplier:   Merck
Papain
Catalog # 1.07144.5000
Supplier:  Merck
CAS Number:  
Papain

—, 5 kg

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 5 kg
Product Details & Documents

Bulk density: 800 kg/m³
Physical form: Fine powder

Documents

Specifications

Specifications

  • Pack type:
  • Pk:
    5 kg
  • Cat. No.:
    1.07144.5000
  • Enzyme activity:
    ≥30000 USP-U/mg (Casein, pH 6,0, 40 °C)
  • Form:
    Yellowish-brown
  • Storage conditions:
    Store below +15 °C
  • pH:
    4 - 7 (1 g/L, H₂O, 25 °C)
  • Enzyme name:
    Papain

Specifications