0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
Orders Link
ContactUsLinkComponent
Buffers
pH Calibration Buffers
pH buffer solutions
pH buffer solutions
:
PanReac AppliChem
pH buffer solutions
:
PanReac AppliChem
pH buffer solutions
APPC272170.1209_TDS_EN
Product Family Options
pH value
Reference temperature
Pk
APPC272170.1209
7,00
20 °C
250 mL
1 * 250 mL
1 * 250 mL - 14,40 €
14,40 €
-
+
1 * 250 mL
14,40 €
-
+
APPC272170.1211
7,00
20 °C
1 L
1 * 1 L
1 * 1 L - 30,50 €
30,50 €
-
+
1 * 1 L
30,50 €
-
+
APPC272170.1315
7,00
20 °C
10 L
1 * 10 L
1 * 10 L - 176,00 €
176,00 €
-
+
1 * 10 L
176,00 €
-
+
Test kits, photometric, AccuVac®
#: 705-0150
: Hach
VWR® Screw Caps for Bottles, Wide Neck
#: 215-5647
: VWR Collection