ContactUsLinkComponent
delta P air flow ComboKit 1 with Bluetooth®, Testo 440
delta P air flow ComboKit 1 with Bluetooth®, Testo 440
Catalog # TEST0563.4409
Supplier:  TESTO
undefined
delta P air flow ComboKit 1 with Bluetooth®, Testo 440
Catalog # TEST0563.4409
Supplier:  TESTO
Specifications
  • Interface:
    Bluetooth
  • Description:
    Air velocity and IAQ measuring instrument, testo 440 delta P Air Flow ComboKit 1 with Bluetooth, differential pressure with internal memory and data export function, universal probe handle with Bluetooth, hot wire probe head incl. temperature and humidity sensor, 100 mm vane probe head incl. temperature sensor, Telescope (1 m) and 90° angle to fit both probes, Combi case for testo 440 dP and multiple probes
  • Seq:
    79
  • Pk:
    1
Specifications
About this item
Delta P air flow ComboKit 1 equipped with clearly structured measurement menu for volume flow and parallel determination of air velocity, differential pressure, humidity and temperature in ventilation ducts or at ventilation outlets.
  • Carry out measurements without any annoying cables
  • With space-saving and integrated differential pressure sensor
  • Easily transported
Convenient with wireless probes, internal data storage and USB port for data export. Ideal for long-term monitoring of indoor air quality and maximum flexibility for duct and outlet measurements.
Delivery information: Supplied with testo 440 dP air velocity and IAQ measuring instrument including differential pressure sensor and the 100 mm vane probe head and hot wire probe head (Ø 9 mm) for attachment to the universal Bluetooth® handle supplied.