ContactUsLinkComponent
Air flow comboKit 1 with Bluetooth®, Testo 440
Air flow comboKit 1 with Bluetooth®, Testo 440
Catalog # TEST0563.4406
Supplier:  TESTO
undefined
Air flow comboKit 1 with Bluetooth®, Testo 440
Catalog # TEST0563.4406
Supplier:  TESTO
Specifications
  • Interface:
    Bluetooth
  • Description:
    Air velocity and IAQ measuring instrument, testo 440 air flow ComboKit 1 with Bluetooth, with internal memory and data export function, 100 mm vane probe with Bluetooth, incl. temperature sensor, Hot wire probe with telescope (0,85 m) incl. temperature sensor, fixed cable (1,8 m), combi case for testo 440 and multiple probes
  • Seq:
    76
  • Pk:
    1
Specifications
About this item
Airflow Combokit 1 equipped with clearly structured measurement menu for volume flow and parallel determination of air velocity, differential pressure, humidity and temperature in ventilation ducts or at ventilation outlets.
  • Long-term monitoring of indoor air quality
  • Combine the vane probe with Bluetooth with the telescope for ceiling outlets
  • Turbulence measurement in accordance with EN ISO 7730 / ASHRAE 55
Convenient for wireless probe, internal data storage and USB port for data export. Ideal for long-term monitoring of indoor air quality and maximum flexibility for duct and outlet measurements.
Delivery information: Supplied with testo 440 air velocity and IAQ measuring instrument, the vane probe (Ø 100 mm) with Bluetooth® including temperature sensor and the hot wire probe including temperature sensor.