Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chromogenic Phosphatase Substrate Sampler kit
Chromogenic Phosphatase Substrate Sampler kit
Chromogenic Phosphatase Substrate Sampler kit
Catalog # 10022.
Supplier:
Biotium
CAS Number:
Expand image
Chromogenic Phosphatase Substrate Sampler kit
Catalog # 10022.
Supplier:
Biotium
CAS Number:
Specifications
Pk:
1 KIT
Seq:
0075
Specifications
About this item
Product Details
:
Storage Temperature:
Refrigerator
Product Details