Order Entry
Belgium
ContactUsLinkComponent
Periodic acid 1% in aqueous solution, Clin-Tech for histology and pathology
Irritant
Periodic acid 1% in aqueous solution, Clin-Tech for histology and pathology
Catalog # VWRK69203/5
Supplier:  KLINIPATH
CAS Number:  
undefined
Irritant
Periodic acid 1% in aqueous solution, Clin-Tech for histology and pathology
Catalog # VWRK69203/5
Supplier:  KLINIPATH
CAS Number:  

Specifications

  • Pk:
    500 ml
  • Seq:
    2725

Specifications

About this item

 :
 :227.94 g/mol
 :GHS07
 :1
CAS Number:10450-60-9
Formula:H₅IO₆
MDL Number:MFCD00011344