Orders LinkContactUsLinkComponent
VWR® Evaporating Flasks
VWR® Evaporating Flasks
Supplier:  VWR Collection
Certificates
VWR® Evaporating Flasks
VWR® Evaporating Flasks
5 Options Available Below
Product Details & Documents
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • White labelling area and imprinted catalogue number
These pear-shaped rotary evaporating flasks are produced from thick walled borosilicate glass tubing. With standard ground socket, suitable for all standard rotary evaporators.
Product Family Options
Product Information