Detection window maker for capillary electrophoresis
About this item
Creates a perfect detection window. Removes the polyimide coating from 365 µm OD capillaries for detection by UV and LIF.
- Easy to use for all capillaries for CE
- Takes 10 seconds to make a perfect window
The Window Maker™ is designed to produce enough heat to burn off the polyimide but not damage the glass in any way. Can be used to remove 1 to 2 mm of the polyimide from the inlet and outlet tips of the capillary.
Delivery information: Supplied with a standard 2 mm module for burning the capillary. Other size heating modules are available as accessories.
3 Options Available Below
Product Details & Documents
Creates a perfect detection window. Removes the polyimide coating from 365 µm OD capillaries for detection by UV and LIF.
- Easy to use for all capillaries for CE
- Takes 10 seconds to make a perfect window
The Window Maker™ is designed to produce enough heat to burn off the polyimide but not damage the glass in any way. Can be used to remove 1 to 2 mm of the polyimide from the inlet and outlet tips of the capillary.
Delivery information: Supplied with a standard 2 mm module for burning the capillary. Other size heating modules are available as accessories.
Product Family Options
Heating module
Product Information
- AvailabilityPrice
- MICS07201-SReplacement heating module, for 2 mm window
- MICS07204-SReplacement heating module, for 4 mm window
- MICS07207-SReplacement heating module, for 7 mm window