Orders LinkContactUsLinkComponent
Universal vials
Universal vials
Universal vials, No
Catalog # EKSIA62251
Supplier:   EKSIGENT
Universal vials
Catalog # EKSIA62251
Supplier:  EKSIGENT
Universal vials
Universal vials, No
Price
132,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Universal vials are used to accommodate buffers/samples.
These polymethylpentene vials are precision manufactured and have been compatibility tested with commonly used CE reagents. Universal vials are designed for use only with CESI 8000, PA 800 Enhanced or PA 800 plus instruments and should not be reused.
Specifications
  • Cap included:
    No
  • Description:
    Universal vials