Order Entry
Belgium
ContactUsLinkComponent
Avantor® Hichrom, HPLC Column Connectors
Avantor® Hichrom, HPLC Column Connectors
Catalog # HICHHI-1091
Supplier:  Avantor
undefined
Avantor® Hichrom, HPLC Column Connectors
Catalog # HICHHI-1091
Supplier:  Avantor

Specifications

  • Description:
    HPLC column connector, 1/16" OD × 0,010" ID
  • Length:
    40 mm
  • Material:
    Stainless steel
  • Pk:
    1

Specifications

About this item

These column connectors are used to connect Hichrom guard and analytical columns.

  • Stainless steel connector
  • Female-female end fittings

The connector consists of a stainless steel tubing with appropriate 1/16" nuts and ferrules and can be used to connect female end fitting Hichrom guard and analytical columns and make similar connections.