Nigrosine 10% staining solution, for microscopy
100 mL
Catalog # 43925-100ML-F
Supplier: SIGMA ALDRICH MICROSCOPY
Nigrosine 10% staining solution, for microscopy
Catalog # 43925-100ML-F
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number: 8005-03-6
Jump to:
Product Details & Documents
Composition: Nigrosin 100 g/l, Formalin 5 ml/l in water
Negative staining method permits visualization of the usually transparent and unstainable capsule of many organisms, most importantly Cryptococcus neoformans.
Synonyms: Acid Black 2
- CAS Number:8005-03-6
- MDL Number:MFCD00044681
- :M-Clarity™ quality level = MQ100
Specifications
Specifications
- Pk:100 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



