Orders LinkContactUsLinkComponent
Verification thermometers
Verification thermometers
Supplier:   Amarell
Verification thermometers
Supplier:  Amarell
Certificates
Verification thermometers
2 Options Available Below
Product Details & Documents
These thermometers are standard set for pharmacy laboratories.
  • Enclosed-scale
  • Milk glass scale
  • Capillary prismatic
  • Richter-top finish with button
It is suitable for temperture range of 0 to 360 °C and has a fine subdivision up to 0,1 °C.
All theremometers in this set are permitted for government verification or calibration.
Product Family Options
Product Information
  • Range
    Length
    Filling
    Immersion depth
    Ø
    Availability
    Price
  • AMARN60180-SET/VC
    Set of verification thermometers with verification certificate
    Set of verification thermometers with verification certificate
    0…+360 °C
    175 mm
    Hg
    Total
    5 - 6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARN60180-SET
    Set of verification thermometers without verification certificate
    Set of verification thermometers without verification certificate
    0…+360 °C
    175 mm
    Hg
    Total
    5 - 6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST