Order Entry
Belgium
ContactUsLinkComponent
Screw caps for laboratory bottles, with temperature probe holder
Screw caps for laboratory bottles, with temperature probe holder
  SCOT299912801
undefined
Screw caps for laboratory bottles, with temperature probe holder
  SCOT299912801

 

  • Thread:
    GL 45
  • Colour:
    Blue
  • Description:
    Screw cap with thermocouple probe holder, Ø (54 mm)
  • Material:
    PTFE
  • Seq:
    01
  • Pk:
    1

 

 

PP caps with stainless steel holder.

  • For use in laboratory autoclaves and sterilisers
  • Maximum temperature 140 °C
  • Autoclavable: 121 °C

The DURAN® temperature probe holder GL 45 consists of a stainless steel holder that is permanently fitted into a blue DURAN® polypropylene GL 45 cap. The holder will accept the 6,0 mm metal temperature measuring probes that are commonly used in laboratory autoclaves and sterilisers.