Order Entry
Belgium
ContactUsLinkComponent
 
Biopharm pressure sensors, SciLog®
Catalog # PARK080-699PSX-5
Supplier:  PARKER HANNIFIN
Biopharm pressure sensors, SciLog®
Catalog # PARK080-699PSX-5
Supplier:  PARKER HANNIFIN

Specifications

  • Maximum Flow Rate:
    1 l/min
  • Range:
    −5...+60 psi
  • Process connection:
    Luer
  • Description:
    Single-use pressure sensor with Luer connection, 60 psi
  • Seq:
    01
  • Wetted materials:
    Animal-Free Compounds
  • Pk:
    1 SET
  • Pipe size:
    ID 0,03" to 0,31"

Specifications

About this item

SciLog® range of pre-calibrated, single-use sensors, can measure and document pressure in a range of filtration, separation and purification applications.

  • Trans-membrane pressure (TMP) control
  • Suitable for single-use GMP applications
  • The sensors are validated for exposure to gamma irradiation, autoclave and sodium hydroxide sterilization

Its applications are growth media preparation, bioreactor maintenance, cell culture harvest, buffer preparation, product filtration, tangential flow filtration, virus control and bulk drug dispense.