ContactUsLinkComponent
Accessories for automatic distillation systems with titration, VAPODEST 50s systems
Accessories for automatic distillation systems with titration, VAPODEST 50s systems
Catalog # GERH12-0354
Supplier:  GERHARDT
Accessories for automatic distillation systems with titration, VAPODEST 50s systems
Catalog # GERH12-0354
Supplier:  GERHARDT
Specifications
  • Description:
    pH-combined electrode KCL, require regularly filling with KCL
  • Seq:
    01
  • Pk:
    1